Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252530.1 | NP_001239459.1 | 6 - 7 | 1109 | 83 | |
NM_001252531.1 | NP_001239460.1 | 6 - 7 | 1052 | 83 | ||
NM_175316.3 | NP_780525.1 | 6 - 7 | 855 | 83 | ||
GenBank mRNA | AK041144.1 | - | 6 - 7 | 1051 | 83 | |
AK048120.1 | - | 6 - 7 | 856 | 83 | ||
AK132394.1 | - | 6 - 7 | 1110 | 83 | ||
AK163542.1 | - | 6 - 7 | 83 | |||
AK173049.1 | - | 4 - 5 | 641 | 83 | ||
BC019209.1 | - | 6 - 7 | 838 | 83 | ||
BC037089.1 | - | 6 - 7 | 827 | 83 | ||
BC049251.1 | - | 6 - 7 | 818 | 83 | ||
BC096485.1 | - | 6 - 7 | 864 | 5101188 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |