Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2183217 |
AK034180.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704988 |
AK019248.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704988 |
XM_484715.4 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704988 |
XM_907118.2 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
715897 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
392444 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
198805 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
45479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607271 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
715684 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
22230 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
352842 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7317 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
162317 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
125974 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102335 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123174 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P269334 |
AK089458.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948858 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948869 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948859 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948871 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948850 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948846 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948853 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948867 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948861 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948855 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948845 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948864 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948862 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948848 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948870 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24948852 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102335 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123174 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06113590 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P269334 |
AK089458.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
BC082583.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
AK078817.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
NM_175372.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P513072 |
AK028434.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P107340 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102335 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123174 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06381924 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06381926 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06143928 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06143925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948792 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948884 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948794 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948842 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948770 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948858 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948755 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948777 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948834 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24948879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948867 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06647835 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06381924 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948846 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06522171 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06301080 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06513428 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948870 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948831 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06143927 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948848 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06421754 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948834 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06302053 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06143926 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948881 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06302050 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948847 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05909624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06301081 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948869 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06421748 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06665139 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948883 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06513430 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06489596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948873 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06302052 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06551481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06381926 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24948871 |
- |
- |