Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177323.3 | NP_796297.2 | 2 - 3 | 179 | 80 | |
GenBank mRNA | AK028707.1 | - | 2 - 3 | 210 | 80 | |
AK133789.1 | - | 2 - 3 | 205 | 80 | ||
AK135334.1 | - | 2 - 3 | 200 | 80 | ||
AK136479.1 | - | 2 - 3 | 185 | 80 | ||
BC065059.1 | - | 2 - 3 | 179 | 6401664 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |