Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177324.2 | NP_796298.2 | 37 - 38 | 5260 | 59 | |
GenBank mRNA | AK029035.1 | - | 4 - 5 | 603 | 59 | |
AK029568.1 | - | 8 - 9 | 1263 | 59 | ||
BC015069.1 | - | 13 - 14 | 1976 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |