Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177381.3 | NP_796355.2 | 8 - 9 | 971 | 112 | |
GenBank mRNA | AK140362.1 | - | 8 - 9 | 971 | 112 | |
BC038030.1 | - | 9 - 10 | 982 | 4020725 | 112 | |
BC048005.1 | - | 4 - 5 | 326 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |