Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500012497.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012497.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012491.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012491.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012492.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012492.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012494.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012494.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012495.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012495.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012498.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012498.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012499.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012499.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012489.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012489.mm.1 |
NM_001040398.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500012493.mm.1 |
TC0500001410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500012493.mm.1 |
NM_001040398.2 |
Refseq |