Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013385.mm.1 |
NM_178389.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013385.mm.1 |
NM_178389.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013385.mm.1 |
TC0400001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013388.mm.1 |
NM_178389.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013388.mm.1 |
NM_178389.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013388.mm.1 |
TC0400001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013386.mm.1 |
NM_178389.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013386.mm.1 |
NM_178389.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013386.mm.1 |
TC0400001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013387.mm.1 |
NM_178389.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013387.mm.1 |
NM_178389.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013387.mm.1 |
TC0400001671.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013390.mm.1 |
NM_178389.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013390.mm.1 |
NM_178389.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013390.mm.1 |
TC0400001671.mm.1 |
MTA TC |