Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178623.3 | NP_848738.2 | 2 - 3 | 108 | 108 | |
GenBank mRNA | AK033577.1 | - | 2 - 3 | 82 | 108 | |
BY058620.1 | - | 2 - 3 | 108 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |