Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfxn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178639.4 | NP_848754.1 | 4 - 5 | 288 | 75 | |
GenBank mRNA | AF325264.1 | - | 4 - 5 | 306 | 75 | |
AK036285.1 | - | 4 - 5 | 286 | 75 | ||
AK043706.1 | - | 4 - 5 | 287 | 75 | ||
AK082073.1 | - | 4 - 5 | 286 | 75 | ||
BB628882.1 | - | 4 - 5 | 289 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Detect Genomic DNA Mus musculus Probe spans exons |