Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161515.1 | NP_001154987.1 | 3 - 4 | 534 | 83 | |
NM_001161516.1 | NP_001154988.1 | 3 - 4 | 315 | 83 | ||
NM_178788.4 | NP_848903.2 | 3 - 4 | 556 | 83 | ||
NR_027759.1 | 3 - 4 | 556 | 83 | |||
GenBank mRNA | AK031645.1 | - | 3 - 4 | 521 | 83 | |
AK039066.1 | - | 3 - 4 | 346 | 83 | ||
AK077663.1 | - | 3 - 4 | 518 | 83 | ||
AK138614.1 | - | 3 - 4 | 344 | 83 | ||
AK171894.1 | - | 3 - 4 | 353 | 83 | ||
BB193146.2 | - | 1 - 2 | 125 | 83 | ||
BC031719.1 | - | 3 - 4 | 340 | 3672932 | 83 | |
CO803572.1 | - | 3 - 4 | 315 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |