Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 7 - 8 | 1117 | 74 | |
NM_001286041.1 | NP_001272970.1 | 6 - 7 | 1054 | 74 | ||
NM_181413.4 | NP_852078.1 | 7 - 8 | 1117 | 74 | ||
GenBank mRNA | AK129094.1 | - | 6 - 7 | 988 | 74 | |
AK147273.1 | - | 7 - 8 | 1052 | 74 | ||
BC050847.1 | - | 7 - 8 | 1083 | 6507130 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |