Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200016895.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016895.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016895.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016895.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016895.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016905.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016905.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016905.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016905.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016905.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016903.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016903.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016903.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016903.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016903.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016899.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016899.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016899.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016899.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016899.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016902.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016902.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016902.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016902.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016902.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016900.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016900.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016900.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016900.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016900.mm.1 |
TC0200002146.mm.1 |
MTA TC |