Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem55a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028264.4 | NP_082540.1 | 2 - 3 | 370 | 121 | |
GenBank mRNA | AK012054.1 | - | 2 - 3 | 330 | 121 | |
AK033845.1 | - | 2 - 3 | 333 | 121 | ||
AK049213.1 | - | 2 - 3 | 371 | 121 | ||
AK206695.1 | - | 2 - 3 | 181 | 121 | ||
AX775959.1 | - | 2 - 3 | 299 | 121 | ||
BC021435.1 | - | 2 - 3 | 315 | 5029766 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |