Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175488.6 | NP_780697.2 | 13 - 14 | 100 | ||
GenBank mRNA | AK029803.1 | - | 13 - 14 | 100 | ||
AK030198.1 | - | 13 - 14 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |