Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175488.6 | NP_780697.2 | 7 - 8 | 810 | 131 | |
GenBank mRNA | AK029803.1 | - | 7 - 8 | 811 | 131 | |
AK030198.1 | - | 7 - 8 | 799 | 131 | ||
AK030205.1 | - | 4 - 5 | 460 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |