Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177265.4 | NP_796239.1 | 1 - 2 | 98 | 109 | |
GenBank mRNA | AK045822.1 | - | 1 - 2 | 99 | 109 | |
AK163573.1 | - | 1 - 2 | 109 | |||
BC049666.1 | - | 1 - 2 | 88 | 6534368 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |