Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177310.2 | NP_796284.1 | 1 - 2 | 99 | 80 | |
GenBank mRNA | AK042110.1 | - | 1 - 2 | 138 | 80 | |
AK088324.1 | - | 1 - 2 | 99 | 80 | ||
BC137569.1 | - | 1 - 2 | 83 | 8860588 | 80 | |
BC144717.1 | - | 1 - 2 | 84 | 9053265 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |