Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168274.1 | NP_001161746.1 | 892 | 119 | ||
NM_177912.4 | NP_808580.2 | 657 | 119 | |||
GenBank mRNA | AK033603.1 | - | 658 | 119 | ||
BC113153.1 | - | 650 | 40058717 | 119 | ||
BC113155.1 | - | 878 | 40058514 | 119 |
Gene Symbol: Gsdmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183194.3 | NP_899017.2 | 5 - 6 | 708 | 119 | |
GenBank mRNA | AK037079.1 | - | 5 - 6 | 709 | 119 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BC099685.1 | - | 752 | Chr. - - [-] on Build | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498798 | AK037079.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P498798 | AK037079.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011101 | NM_177912.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011101 | AK033603.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011101 | NM_001168274.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011101 | NM_183194.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2011101 | AK037079.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763123 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481762 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779818 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768749 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674211 | AF235001.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674211 | AF380353.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674211 | AK046718.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674211 | NM_053248.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555108 | AK016545.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 161124 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 124218 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139906 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011101 | NM_177912.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011101 | AK033603.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011101 | NM_001168274.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011101 | NM_183194.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2011101 | AK037079.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498798 | AK037079.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P498798 | AK037079.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139906 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03053126 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08393536 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08393417 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08393462 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03053113 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08393460 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03053122 | - | - |
Whole Mouse Genome G4122A | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498798 | AK037079.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498798 | NM_183194.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498798 | AK037079.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P87997 | BC113155.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P87997 | NM_177912.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P87997 | AK033603.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P87997 | BC113153.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P87997 | NM_001168274.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134624 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104052 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138990 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03053122 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03053113 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08393536 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139906 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08393462 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08393417 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30277625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30277662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30277636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30277659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30277660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277670 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12097717 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12097734 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30277648 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10428975 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428982 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428957 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428962 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428974 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428971 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428968 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428965 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428969 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428961 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428972 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428967 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428964 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428966 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428956 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428958 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428970 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428963 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10428959 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10428973 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10428960 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10428955 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10428981 | - | - |
Mouse Genome 430 2.0 Array | 1454883_at | - | - |
Mouse Genome 430 2.0 Array | 1454883_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 331063_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 331063_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500012127.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012127.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012127.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012127.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012127.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012128.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012128.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012128.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012128.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012128.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012165.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012165.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012165.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012159.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012159.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012159.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012160.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012160.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012160.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012129.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012129.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012129.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012129.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012129.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012158.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012158.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012158.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012161.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012161.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012161.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012164.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012164.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012164.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012121.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012121.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012121.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012121.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012121.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012125.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012125.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012125.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012125.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012125.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012126.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012126.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012126.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012126.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012126.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012120.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012120.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012120.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012120.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012120.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012123.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012123.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012123.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012123.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012123.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012162.mm.1 | NM_183194.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012162.mm.1 | NM_183194.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012162.mm.1 | TC1500001622.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012122.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012122.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012122.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012122.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012122.mm.1 | TC1500002344.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500012124.mm.1 | NM_001168274.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012124.mm.1 | NM_001168274.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012124.mm.1 | NM_177912.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012124.mm.1 | NM_177912.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500012124.mm.1 | TC1500002344.mm.1 | MTA TC |
Mouse Expression Set 430 | 1430641_PM_at | - | - |
Mouse Expression Set 430 | 1430641_at | - | - |
Mouse Expression Set 430 | 1454883_at | - | - |
Mouse Expression Set 430 | 1454883_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 362843 | - | - |
ABI Mouse Genome Survey Microarray | 522239 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4540022 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104730059 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4540022 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103440347 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2900020 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004540022 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900020 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2672969 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753714 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2753714 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1246145 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2672969 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214305 | - | - |
mouse-6 v1.1 (Illumina) | GI_34147259-S | - | - |
mouse-6 v1.1 (Illumina) | scl00331063.1_84-S | - | - |
mouse-6 v1.1 (Illumina) | scl00270328.1_149-S | - | - |
mouse-6 v1.1 (Illumina) | GI_31342689-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900020 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4540022 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103440347 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104730059 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00331063.1_84-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00270328.1_149-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103440347 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900020 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104730059 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4540022 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3115255 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2753714 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3040543 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2672969 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3040543 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246145 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2753714 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2672969 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1214305 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3115255 | - | - |