Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178036.4 | NP_828875.1 | 2 - 3 | 302 | 80 | |
GenBank mRNA | AF435737.1 | - | 2 - 3 | 302 | 80 | |
AK020338.1 | - | 1 - 2 | 50 | 80 | ||
BC119103.1 | - | 2 - 3 | 270 | 8733852 | 80 | |
BC120657.1 | - | 2 - 3 | 270 | 40129580 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |