Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037136.1 | NP_001032213.1 | 14 - 15 | 2355 | 60 | |
NM_178119.3 | NP_835220.1 | 15 - 16 | 2514 | 60 | ||
GenBank mRNA | AK035494.1 | - | 4 - 5 | 568 | 60 | |
AK046523.1 | - | 15 - 16 | 2475 | 60 | ||
AK129289.2 | - | 15 - 16 | 2461 | 60 | ||
AK147503.1 | - | 15 - 16 | 2520 | 60 | ||
AK147521.1 | - | 15 - 16 | 3006 | 60 | ||
AK213954.1 | - | 3 - 4 | 348 | 60 | ||
BC096588.1 | - | 14 - 15 | 2303 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |