Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038710.1 | NP_001033799.1 | 3 - 4 | 714 | 76 | |
NM_016711.3 | NP_057920.2 | 3 - 4 | 650 | 76 | ||
GenBank mRNA | AK015719.1 | - | 3 - 4 | 750 | 76 | |
AK017072.1 | - | 3 - 4 | 696 | 77 | ||
AK018223.1 | - | 3 - 4 | 447 | 76 | ||
AK076562.1 | - | 3 - 4 | 664 | 76 | ||
AK079077.1 | - | 3 - 4 | 494 | 76 | ||
AK164178.1 | - | 3 - 4 | 496 | 76 | ||
BC052227.1 | - | 3 - 4 | 714 | 76 | ||
BC061124.1 | - | 3 - 4 | 480 | 6531518 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |