Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016796.3 | NP_058076.2 | 5 - 6 | 365 | 64 | |
GenBank mRNA | AK002977.1 | - | 5 - 6 | 367 | 64 | |
AK011678.1 | - | 5 - 6 | 345 | 64 | ||
AK018344.1 | - | 5 - 6 | 376 | 64 | ||
AK031151.1 | - | 5 - 6 | 367 | 64 | ||
BC138753.1 | - | 4 - 5 | 320 | 8861775 | 64 | |
BC138754.1 | - | 4 - 5 | 320 | 8861776 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |