Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK167383.1 | - | 7 - 8 | 787 | 120 | |
AK167848.1 | - | 7 - 8 | 783 | 120 | ||
AY082332.1 | - | 7 - 8 | 753 | 120 | ||
BC050206.1 | - | 7 - 8 | 761 | 30009463 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |