Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153788.3 | NP_722483.2 | 4 - 5 | 490 | 68 | |
GenBank mRNA | AK133473.1 | - | 3 - 4 | 714 | 68 | |
AK138736.1 | - | 4 - 5 | 482 | 68 | ||
AK154210.1 | - | 4 - 5 | 455 | 68 | ||
AK154449.1 | - | 4 - 5 | 493 | 68 | ||
AK155001.1 | - | 4 - 5 | 511 | 68 | ||
BC031462.1 | - | 4 - 5 | 490 | 4017986 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |