Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110251.1 | NP_001103721.1 | 3 - 4 | 280 | 112 | |
NM_013551.2 | NP_038579.2 | 3 - 4 | 338 | 112 | ||
GenBank mRNA | AK028234.1 | - | 3 - 4 | 205 | 112 | |
AK143558.1 | - | 3 - 4 | 205 | 112 | ||
AK151020.1 | - | 3 - 4 | 304 | 112 | ||
AK166734.1 | - | 3 - 4 | 236 | 112 | ||
AK167702.1 | - | 3 - 4 | 297 | 112 | ||
BB844509.1 | - | 3 - 4 | 281 | 112 | ||
BC003861.1 | - | 3 - 4 | 298 | 3487642 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Mus musculus Mouse Endogenous Controls Inventoried Mouse_Endogenous_Controls Probe spans exons |