Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027984.3 | NP_082260.1 | 8 - 9 | 1771 | 106 | |
GenBank mRNA | AK009469.1 | - | 7 - 8 | 1151 | 106 | |
AK158031.1 | - | 7 - 8 | 1649 | 106 | ||
BC016454.1 | - | 8 - 9 | 1720 | 4217221 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |