Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028762.1 | NP_083038.1 | 1 - 2 | 138 | 62 | |
GenBank mRNA | AK053511.1 | - | 1 - 2 | 142 | 62 | |
AK146493.1 | - | 1 - 2 | 168 | 62 | ||
AK167100.1 | - | 1 - 2 | 134 | 62 | ||
BC025619.1 | - | 1 - 2 | 138 | 5320947 | 62 | |
BC034010.1 | - | 1 - 2 | 104 | 4225239 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |