Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168403.1 | NP_001161875.1 | 7 - 8 | 1193 | 99 | |
NM_001168404.1 | NP_001161876.1 | 7 - 8 | 730 | 99 | ||
NM_029898.3 | NP_084174.2 | 6 - 7 | 1087 | 99 | ||
GenBank mRNA | AK045480.1 | - | 6 - 7 | 721 | 99 | |
AK081197.1 | - | 7 - 8 | 731 | 99 | ||
AK138273.1 | - | 6 - 7 | 742 | 99 | ||
AK140417.1 | - | 7 - 8 | 1057 | 99 | ||
BC152317.1 | - | 6 - 7 | 714 | 40088531 | 99 | |
BC152536.1 | - | 6 - 7 | 714 | 40088530 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |