Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008784.3 | NP_032810.4 | 2 - 3 | 566 | 89 | |
GenBank mRNA | AA213189.1 | - | 1 - 2 | 93 | 89 | |
AK017166.1 | - | 2 - 3 | 490 | 89 | ||
BC089567.1 | - | 2 - 3 | 498 | 30297519 | 89 | |
BU520087.1 | - | 2 - 3 | 504 | 89 | ||
CX203560.1 | - | 2 - 3 | 481 | 89 | ||
DV654442.1 | - | 2 - 3 | 601 | 89 | ||
L31652.1 | - | 2 - 3 | 491 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |