Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009855.2 | NP_033985.3 | 2 - 3 | 676 | 117 | |
GenBank mRNA | AF065893.1 | - | 2 - 3 | 485 | 117 | |
AF065894.1 | - | 2 - 3 | 485 | 117 | ||
AF065895.1 | - | 2 - 3 | 485 | 117 | ||
AF065896.1 | - | 2 - 3 | 485 | 117 | ||
AJ278965.1 | - | 2 - 3 | 428 | 117 | ||
AK154329.1 | - | 3 - 4 | 785 | 117 | ||
AY278186.1 | - | 2 - 3 | 476 | 117 | ||
BC131959.1 | - | 2 - 3 | 496 | 40130236 | 117 | |
BC145843.1 | - | 2 - 3 | 496 | 40131146 | 117 | |
X60958.1 | - | 2 - 3 | 676 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Immune Response 1700 Inventoried Mouse_Immune_Response Probe spans exons |