Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009445.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009445.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009445.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009460.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009460.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009460.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009459.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009459.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009459.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009446.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009446.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009446.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009447.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009447.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009447.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009448.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009448.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009448.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009466.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009466.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009466.mm.1 |
NM_028385.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009465.mm.1 |
TC0600001239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009465.mm.1 |
NM_028385.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009465.mm.1 |
NM_028385.1 |
Refseq |