Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028976.2 | NP_083252.1 | 2 - 3 | 177 | 67 | |
GenBank mRNA | AK154986.1 | - | 2 - 3 | 154 | 67 | |
AK170627.1 | - | 2 - 3 | 141 | 67 | ||
BC012251.1 | - | 2 - 3 | 177 | 4166619 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |