Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173865.3 | NP_776290.1 | 9 - 10 | 2200 | 72 | |
GenBank mRNA | AK089802.1 | - | 9 - 10 | 2201 | 72 | |
AK154819.1 | - | 9 - 10 | 2175 | 72 | ||
BC037033.1 | - | 4 - 5 | 509 | 72 | ||
BC096596.1 | - | 9 - 10 | 2236 | 4239523 | 72 | |
BC116280.1 | - | 9 - 10 | 2008 | 40094408 | 72 | |
BC116281.1 | - | 9 - 10 | 2008 | 40094411 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |