Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173867.5 | NP_776292.1 | 9 - 10 | 1389 | 73 | |
GenBank mRNA | AK076040.1 | - | 9 - 10 | 1320 | 73 | |
AK076046.1 | - | 1620 | 73 | |||
AK129370.1 | - | 8 - 9 | 1093 | 73 | ||
BC020385.1 | - | 7 - 8 | 922 | 73 | ||
BC034759.1 | - | 6 - 7 | 709 | 73 | ||
BC060624.1 | - | 9 - 10 | 1343 | 73 | ||
BC086666.1 | - | 9 - 10 | 1312 | 30634817 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |