Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175214.3 | NP_780423.2 | 6 - 7 | 1976 | 92 | |
GenBank mRNA | BC116646.1 | - | 4 - 5 | 815 | 40086396 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |