Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_001161432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_001161432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_001161433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_001161433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_175540.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
NM_175540.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016842.mm.1 |
TC0X00002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_001161432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_001161432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_001161433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_001161433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_175540.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
NM_175540.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016847.mm.1 |
TC0X00002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_001161432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_001161432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_001161433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_001161433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_175540.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
NM_175540.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016845.mm.1 |
TC0X00002685.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_001161432.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_001161432.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_001161433.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_001161433.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_175540.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
NM_175540.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00016843.mm.1 |
TC0X00002685.mm.1 |
MTA TC |