Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242349.1 | NP_001229278.1 | 11 - 12 | 1370 | 76 | |
NM_178642.5 | NP_848757.4 | 11 - 12 | 1367 | 76 | ||
GenBank mRNA | AK032793.1 | - | 6 - 7 | 378 | 76 | |
AK052589.1 | - | 11 - 12 | 1319 | 76 | ||
BC004697.1 | - | 3 - 4 | 233 | 76 | ||
BC062959.1 | - | 11 - 12 | 1256 | 30547439 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |