Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200007665.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007665.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007672.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007672.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007673.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007673.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007666.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007666.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007668.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007668.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007680.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007680.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007664.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007664.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007675.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007675.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007662.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007662.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007663.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007663.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007671.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007671.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007677.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007677.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007682.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007682.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007667.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007667.mm.1 |
TC1200001072.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007669.mm.1 |
NM_011112.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007669.mm.1 |
TC1200001072.mm.1 |
MTA TC |