Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009697.3 | NP_033827.2 | 2 - 3 | 1232 | 65 | |
NM_183261.3 | NP_899084.2 | 2 - 3 | 1180 | 65 | ||
GenBank mRNA | AK034619.1 | - | 1 - 2 | 351 | 65 | |
AK140125.1 | - | 2 - 3 | 1156 | 65 | ||
BC042484.1 | - | 2 - 3 | 1232 | 5368085 | 65 | |
BC094360.1 | - | 2 - 3 | 1157 | 65 | ||
U07635.1 | - | 2 - 3 | 1115 | 65 | ||
X76653.1 | - | 2 - 3 | 1003 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |