Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncald | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170866.1 | NP_001164337.1 | 5 - 6 | 675 | 155 | |
NM_001170867.1 | NP_001164338.1 | 3 - 4 | 609 | 155 | ||
NM_001170868.1 | NP_001164339.1 | 2 - 3 | 557 | 155 | ||
NM_134094.4 | NP_598855.2 | 5 - 6 | 865 | 155 | ||
GenBank mRNA | AK034031.1 | - | 3 - 4 | 588 | 155 | |
AK048440.1 | - | 5 - 6 | 845 | 155 | ||
AK167313.1 | - | 2 - 3 | 527 | 155 | ||
BC011162.1 | - | 2 - 3 | 552 | 4191195 | 155 | |
BC026979.1 | - | 2 - 3 | 511 | 5365560 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |