Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516076 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516080 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515850 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515849 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516075 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515845 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515847 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516078 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515848 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516079 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515846 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013465 |
AK168429.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013465 |
BC016226.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013465 |
AK145228.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013465 |
AK152450.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2013465 |
NM_145443.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2097863 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
606422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
231884 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563669 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170042 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
713741 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
838787 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
658462 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
953457 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186296 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26861 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277327 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
221831 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855448 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826065 |
AK040982.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826065 |
AK041165.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826065 |
AK170671.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826065 |
NM_178792.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
291002 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
919257 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593290 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
240022 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
276195 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153139 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
116479 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84954 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
160896 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
118084 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P175817 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P153578 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2097863 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013465 |
AK168429.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013465 |
BC016226.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013465 |
AK145228.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013465 |
AK152450.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2013465 |
NM_145443.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516070 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516060 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516079 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516063 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516080 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516077 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516074 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516073 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28516068 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P153578 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07655088 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07655051 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
AK168429.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
BC016226.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
NM_145443.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P221132 |
AK139539.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P153578 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09955598 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09955597 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09955600 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09955595 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516076 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515846 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515847 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515845 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516080 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516075 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516078 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515849 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515848 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28515850 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28516079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516077 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515909 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516079 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516063 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515952 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515967 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515882 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515887 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515924 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515953 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515972 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515961 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515975 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515921 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515933 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515962 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515874 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28516061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515869 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515938 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515944 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515935 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515917 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515978 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515956 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515854 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515929 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515963 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515974 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28515958 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515871 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515863 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515844 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09825537 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516064 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515869 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515875 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516068 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516073 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515859 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515852 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515887 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515850 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515853 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515874 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10424587 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515856 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515873 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515867 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515840 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09888802 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515883 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516077 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516063 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516074 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P09888795 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515876 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515884 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10234710 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516071 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515841 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515851 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515864 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515854 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515861 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516061 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515889 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28515857 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28516069 |
- |
- |