Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282021.1 | NP_001268950.1 | 3 - 4 | 679 | 144 | |
NM_009499.3 | NP_033525.2 | 3 - 4 | 679 | 144 | ||
GenBank mRNA | AF084548.1 | - | 3 - 4 | 646 | 144 | |
AK140329.1 | - | 3 - 4 | 643 | 144 | ||
AK150103.1 | - | 3 - 4 | 641 | 144 | ||
AK155480.1 | - | 3 - 4 | 641 | 145 | ||
AK157021.1 | - | 3 - 4 | 643 | 144 | ||
AK171302.1 | - | 3 - 4 | 643 | 145 | ||
AK171715.1 | - | 3 - 4 | 643 | 144 | ||
BC010223.1 | - | 3 - 4 | 381 | 3500587 | 144 | |
BC015289.1 | - | 3 - 4 | 622 | 4240907 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |