Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010019.3 | NP_034149.2 | 9 - 10 | 983 | 68 | |
GenBank mRNA | AB018002.1 | - | 9 - 10 | 973 | 68 | |
AF052942.1 | - | 7 - 8 | 663 | 68 | ||
BC022165.1 | - | 9 - 10 | 952 | 4016911 | 68 | |
BX527735.1 | - | 5 - 6 | 367 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |