Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010713.2 | NP_034843.2 | 3 - 4 | 539 | 69 | |
GenBank mRNA | AJ000338.1 | - | 3 - 4 | 639 | 69 | |
AK160121.1 | - | 3 - 4 | 540 | 69 | ||
AK165733.1 | - | 3 - 4 | 533 | 69 | ||
BC125281.1 | - | 3 - 4 | 448 | 40129796 | 69 | |
BC125283.1 | - | 3 - 4 | 448 | 40129798 | 69 | |
BC144768.1 | - | 3 - 4 | 448 | 9053316 | 69 | |
D49658.1 | - | 3 - 4 | 508 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |