Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011447.3 | NP_035577.1 | 1 - 2 | 579 | 64 | |
GenBank mRNA | AK032459.1 | - | 1 - 2 | 580 | 64 | |
AK033460.1 | - | 1 - 2 | 580 | 64 | ||
AK034231.1 | - | 1 - 2 | 580 | 64 | ||
AK046473.1 | - | 1 - 2 | 580 | 64 | ||
AK053716.1 | - | 1 - 2 | 580 | 64 | ||
AK145294.1 | - | 1 - 2 | 580 | 64 | ||
BC085619.1 | - | 1 - 2 | 555 | 6306276 | 64 | |
Z18957.1 | - | 1 - 2 | 98 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |