Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012055.3 | NP_036185.1 | 7 - 8 | 1127 | 109 | |
GenBank mRNA | AK076207.1 | - | 7 - 8 | 1145 | 109 | |
AK160459.1 | - | 7 - 8 | 1142 | 109 | ||
AK167524.1 | - | 7 - 8 | 1107 | 109 | ||
AK199543.1 | - | 3 - 4 | 324 | 109 | ||
BC005552.1 | - | 7 - 8 | 1125 | 3585775 | 109 | |
BC050123.1 | - | 3455 | 109 | |||
U38940.1 | - | 7 - 8 | 1128 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |