Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk32b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022416.2 | NP_071861.1 | 6 - 7 | 933 | 65 | |
GenBank mRNA | AJ250840.1 | - | 6 - 7 | 1011 | 65 | |
BC052404.1 | - | 6 - 7 | 843 | 5715729 | 65 | |
BC056396.1 | - | 6 - 7 | 919 | 6841272 | 65 | |
BC058412.1 | - | 6 - 7 | 843 | 5715729 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |