Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028243.3 | NP_082519.1 | 1 - 2 | 249 | 96 | |
GenBank mRNA | AK011112.1 | - | 1 - 2 | 231 | 96 | |
AK011816.1 | - | 1 - 2 | 203 | 96 | ||
AK051677.1 | - | 1 - 2 | 206 | 96 | ||
AK054462.1 | - | 1 - 2 | 187 | 96 | ||
AK166282.1 | - | 1 - 2 | 206 | 96 | ||
BC026424.1 | - | 1 - 2 | 45 | 96 | ||
BC055022.1 | - | 1 - 2 | 182 | 4017462 | 96 | |
BC089575.1 | - | 1 - 2 | 204 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |