Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008014.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008014.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008021.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008021.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008025.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008025.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008015.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008015.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008029.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008029.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008020.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008020.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008024.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008024.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008023.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008023.mm.1 |
TC1900000916.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900008027.mm.1 |
NM_130863.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008027.mm.1 |
TC1900000916.mm.1 |
MTA TC |