Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181397.2 | NP_852062.1 | 4 - 5 | 730 | 69 | |
GenBank mRNA | AK082517.1 | - | 343 | 69 | ||
AK087803.1 | - | 503 | 69 | |||
AK088466.1 | - | 2 - 3 | 267 | 69 | ||
AK152130.1 | - | 4 - 5 | 652 | 69 | ||
AK153288.1 | - | 4 - 5 | 652 | 69 | ||
AK155870.1 | - | 4 - 5 | 694 | 69 | ||
AK170913.1 | - | 4 - 5 | 693 | 69 | ||
AK172884.1 | - | 4 - 5 | 658 | 69 | ||
BC052074.1 | - | 4 - 5 | 730 | 6401192 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |